Anti-EIF2G Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-53087
102183-630EA 564.74 USD
102183-630
Anti-EIF2G Rabbit Polyclonal Antibody
Antibodies
The EIF2G Antibody from Novus Biologicals is a rabbit polyclonal antibody to EIF2G. This antibody reacts with human. The EIF2G Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: EIF2G
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR