Anti-EIF1AX Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-80556
102216-758EA 564.74 USD
102216-758
Anti-EIF1AX Rabbit Polyclonal Antibody
Antibodies
The EIF1AX Antibody from Novus Biologicals is a rabbit polyclonal antibody to EIF1AX. This antibody reacts with human. The EIF1AX Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: EIF1AX
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR