Anti-EHD4 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-54873
102184-706EA 564.74 USD
102184-706
Anti-EHD4 Rabbit Polyclonal Antibody
Antibodies
The EHD4 Antibody from Novus Biologicals is a rabbit polyclonal antibody to EHD4. This antibody reacts with human. The EHD4 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: EHD4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR