Anti-EGR2 Rabbit Polyclonal Antibody (DyLight 650)

Supplier: Novus Biologicals
NB110-59723C
102123-846EA 614.15 USD
102123-846
Anti-EGR2 Rabbit Polyclonal Antibody (DyLight 650)
Antibodies
The EGR2 Antibody [DyLight 650] from Novus Biologicals is a rabbit polyclonal antibody to EGR2. This antibody reacts with human, mouse, porcine. The EGR2 Antibody [DyLight 650] has been validated for the following applications: Western Blot, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: EGR2
Clonality: Polyclonal
Clone:
Conjugation: DyLight 650
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Porcine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR