Anti-EGR1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-02807
102155-550EA 429 USD
102155-550
Anti-EGR1 Rabbit Polyclonal Antibody
Antibodies
The EGR1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to EGR1. This antibody reacts with human. The EGR1 Antibody has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Recommended Dilutions: Immunohistochemistry (Parafin): 10 ?g/ml

Type: Primary
Antigen: EGR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR