Anti-EGF Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-19806
102159-530EA 329 USD
102159-530
Anti-EGF Rabbit Polyclonal Antibody
Antibodies
The EGF Antibody from Novus Biologicals is a rabbit polyclonal antibody to EGF. This antibody reacts with human, mouse, rat. The EGF Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:50-1:200

Type: Primary
Antigen: EGF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR