Anti-Eg5 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB500-181 NB500-181SS
102131-936EA 208 USD
102131-936 102131-934
Anti-Eg5 Rabbit Polyclonal Antibody
Antibodies
The Eg5 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Eg5. This antibody reacts with human, mouse, rat, porcine, drosophila. The Eg5 Antibody has been validated for the following applications: Western Blot, Immunocytochemistry / Immunofluorescence, Immunoprecipitation.

Type: Primary
Antigen: Eg5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Porcine, Drosophila
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR