Anti-EFHD1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-71995
102201-786EA 399 USD
102201-786
Anti-EFHD1 Rabbit Polyclonal Antibody
Antibodies
The EFHD1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to EFHD1. This antibody reacts with human, mouse, rat. The EFHD1 Antibody has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry-Paraffin, Immunofluorescence.

Recommended Dilutions: Immunohistochemistry (Parafin): 5 ?g/ml

Type: Primary
Antigen: EFHD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR