Anti-EFCAB4B Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-76493
102208-232EA 564.74 USD
102208-232
Anti-EFCAB4B Rabbit Polyclonal Antibody
Antibodies
The EFCAB4B Antibody from Novus Biologicals is a rabbit polyclonal antibody to EFCAB4B. This antibody reacts with human, mouse. The EFCAB4B Antibody has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: EFCAB4B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR