Anti-EEF2K Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-78959
102213-546EA 379 USD
102213-546
Anti-EEF2K Rabbit Polyclonal Antibody
The EEF2K [p Ser366] Antibody from Novus Biologicals is a rabbit polyclonal antibody to EEF2K. This antibody reacts with human, rat, primate. The EEF2K [p Ser366] Antibody has been validated for the following applications: Western Blot, Immunoprecipitation.

Type: Primary
Antigen: EEF2K
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat, Primate
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR