Anti-EEF1E1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-32184
102165-604EA 635.51 USD
102165-604
Anti-EEF1E1 Rabbit Polyclonal Antibody
Antibodies
The EEF1E1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to EEF1E1. This antibody reacts with human, mouse. The EEF1E1 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: EEF1E1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR