Anti-EEA1 Rabbit Polyclonal Antibody

NBP1-05961
102156-960EA 536.92 USD
102156-960
Anti-EEA1 Rabbit Polyclonal Antibody
The EEA1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to EEA1. This antibody reacts with human. The EEA1 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: EEA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR