Anti-EDA2R Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-76710
102209-066EA 564.74 USD
102209-066
Anti-EDA2R Rabbit Polyclonal Antibody
Antibodies
The EDA2R / TNFRSF27 / XEDAR Antibody from Novus Biologicals is a rabbit polyclonal antibody to EDA2R / TNFRSF27 / XEDAR. This antibody reacts with human. The EDA2R / TNFRSF27 / XEDAR Antibody has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: EDA2R
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR