Anti-EDA Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-80536
102216-718EA 564.74 USD
102216-718
Anti-EDA Rabbit Polyclonal Antibody
Antibodies
The EDA / Ectodysplasin Antibody from Novus Biologicals is a rabbit polyclonal antibody to EDA / Ectodysplasin. This antibody reacts with human. The EDA / Ectodysplasin Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: EDA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR