Anti-ECSIT Rabbit Polyclonal Antibody

NBP1-77838
102212-106EA 536.92 USD
102212-106
Anti-ECSIT Rabbit Polyclonal Antibody
The ECSIT Antibody from Novus Biologicals is a rabbit polyclonal antibody to ECSIT. This antibody reacts with human, mouse. The ECSIT Antibody has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: ECSIT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR