Anti-PA2G4/EBP1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-71840
102201-554EA 635.51 USD
102201-554
Anti-PA2G4/EBP1 Rabbit Polyclonal Antibody
Antibodies
The PA2G4 / EBP1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to PA2G4 / EBP1. This antibody reacts with human, mouse. The PA2G4 / EBP1 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: PA2G4/EBP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR