Anti-EBI2/GPR183 Rabbit Polyclonal Antibody

NBP1-44160
102173-886EA 493.85 USD
102173-886
Anti-EBI2/GPR183 Rabbit Polyclonal Antibody
The EBI2 / GPR183 Antibody from Novus Biologicals is a rabbit polyclonal antibody to EBI2 / GPR183. This antibody reacts with human. The EBI2 / GPR183 Antibody has been validated for the following applications: Immunohistochemistry-Paraffin.

Type: Primary
Antigen: EBI2/GPR183
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR