Anti-E4F1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB100-2871
102108-280EA 635.51 USD
102108-280
Anti-E4F1 Rabbit Polyclonal Antibody
Antibodies
The E4F1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to E4F1. This antibody reacts with human. The E4F1 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: E4F1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR