Anti-E2F-1 Rabbit Polyclonal Antibody

NB100-1629
102105-672EA 395.38 USD
102105-672
Anti-E2F-1 Rabbit Polyclonal Antibody
The E2F-1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to E2F-1. This antibody reacts with human. The E2F-1 Antibody has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen.

Type: Primary
Antigen: E2F-1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR