Anti-E. Coli Rabbit Polyclonal Antibody (Biotin)

Supplier: Novus Biologicals
NB100-62528
102111-776EA 493 USD
102111-776
Anti-E. Coli Rabbit Polyclonal Antibody (Biotin)
Antibodies
The E. Coli Antibody [Biotin] from Novus Biologicals is a rabbit polyclonal antibody to E. Coli. This antibody reacts with bacteria. The E. Coli Antibody [Biotin] has been validated for the following applications: ELISA.

Type: Primary
Antigen: E. Coli
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bacteria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR