Anti-DNM Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB300-565
102130-822EA 564.74 USD
102130-822
Anti-DNM Rabbit Polyclonal Antibody
Antibodies
The Dynamin Antibody from Novus Biologicals is a rabbit polyclonal antibody to Dynamin. This antibody reacts with mouse, rat. The Dynamin Antibody has been validated for the following applications: Western Blot, Immunocytochemistry.

Type: Primary
Antigen: DNM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR