Anti-DUSP26 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-31254
102164-364EA 635.51 USD
102164-364
Anti-DUSP26 Rabbit Polyclonal Antibody
Antibodies
The DUSP26 Antibody from Novus Biologicals is a rabbit polyclonal antibody to DUSP26. This antibody reacts with human, mouse. The DUSP26 Antibody has been validated for the following applications: Western Blot, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: DUSP26
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR