Anti-DUSP10 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-31001
102163-978EA 635.51 USD
102163-978
Anti-DUSP10 Rabbit Polyclonal Antibody
Antibodies
The DUSP10 Antibody from Novus Biologicals is a rabbit polyclonal antibody to DUSP10. This antibody reacts with human. The DUSP10 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: DUSP10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR