Anti-CHIP/STUB1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB100-77315
102116-896EA 635.51 USD
102116-896
Anti-CHIP/STUB1 Rabbit Polyclonal Antibody
Antibodies
The CHIP / STUB1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to CHIP / STUB1. This antibody reacts with human, mouse. The CHIP / STUB1 Antibody has been validated for the following applications: Western Blot, Immunoprecipitation, Proximity Ligation Assay.

Type: Primary
Antigen: CHIP/STUB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR