Anti-Beclin 2 Rabbit Polyclonal Antibody (DyLight 650)

Supplier: Novus Biologicals
NB110-60984C
102124-354EA 614.15 USD
102124-354
Anti-Beclin 2 Rabbit Polyclonal Antibody (DyLight 650)
Antibodies
The Beclin 2 Antibody [DyLight 650] from Novus Biologicals is a rabbit polyclonal antibody to Beclin 2. This antibody reacts with human. The Beclin 2 Antibody [DyLight 650] has been validated for the following applications: Western Blot, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: Beclin 2
Clonality: Polyclonal
Clone:
Conjugation: DyLight 650
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR