Anti-BCR Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-19097
102158-496EA 649.66 USD
102158-496
Anti-BCR Rabbit Polyclonal Antibody
Antibodies
The BCR Antibody from Novus Biologicals is a rabbit polyclonal antibody to BCR. This antibody reacts with human. The BCR Antibody has been validated for the following applications: Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Proximity Ligation Assay.

Proximity Ligation Assay

Type: Primary
Antigen: BCR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR