Anti-BCMA/TNFRSF17 Rabbit Polyclonal Antibody

NBP1-50674
102180-464EA 536.92 USD
102180-464
Anti-BCMA/TNFRSF17 Rabbit Polyclonal Antibody
The BCMA / TNFRSF17 Antibody from Novus Biologicals is a rabbit polyclonal antibody to BCMA / TNFRSF17. This antibody reacts with human. The BCMA / TNFRSF17 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: BCMA/TNFRSF17
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR