Anti-Bcl-xL Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB110-83982 NB110-83982SS
102125-930EA 564.74 USD
102125-930 102125-932
Anti-Bcl-xL Rabbit Polyclonal Antibody
Antibodies
The Bcl-xL Antibody from Novus Biologicals is a rabbit polyclonal antibody to Bcl-xL. This antibody reacts with human, canine, feline, sheep. The Bcl-xL Antibody has been validated for the following applications: Western Blot, ICC / IF (Negative).

Type: Primary
Antigen: Bcl-xL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Canine, Feline, Sheep
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR