Anti-BCL11B Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB600-263
102133-654EA 635.51 USD
102133-654
Anti-BCL11B Rabbit Polyclonal Antibody
Antibodies
The BCL11B Antibody from Novus Biologicals is a rabbit polyclonal antibody to BCL11B. This antibody reacts with human. The BCL11B Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: BCL11B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR