Anti-Bag-1 Rabbit Polyclonal Antibody

NBP1-76714
102209-080EA 466.15 USD
102209-080
Anti-Bag-1 Rabbit Polyclonal Antibody
The Bag-1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Bag-1. This antibody reacts with human, mouse, rat. The Bag-1 Antibody has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: Bag-1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR