Anti-BACE-1 Rabbit Polyclonal Antibody

NBP1-72126
102201-958EA 536.92 USD
102201-958
Anti-BACE-1 Rabbit Polyclonal Antibody
The BACE-1 [p Ser498] Antibody from Novus Biologicals is a rabbit polyclonal antibody to BACE-1. This antibody reacts with human, mouse. The BACE-1 [p Ser498] Antibody has been validated for the following applications: Western Blot, Immunohistochemistry-Paraffin.

  • Recommended Dilutions: Immunohistochemistry (Parafin): 5 μg/ml

Type: Primary
Antigen: BACE-1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR