Anti-Baboon Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB300-664
102130-942EA 805.35 USD
102130-942
Anti-Baboon Rabbit Polyclonal Antibody
Antibodies
The Baboon Antibody from Novus Biologicals is a rabbit polyclonal antibody to Baboon. This antibody reacts with drosophila. The Baboon Antibody has been validated for the following applications: Western Blot, Immunocytochemistry / Immunofluorescence, Immunoprecipitation.

Type: Primary
Antigen: Baboon
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Drosophila
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR