Anti-AXUD1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB200-144
102129-132EA 279 USD
102129-132
Anti-AXUD1 Rabbit Polyclonal Antibody
Antibodies
The AXUD1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to AXUD1. This antibody reacts with human. The AXUD1 Antibody has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Type: Primary
Antigen: AXUD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR