Anti-Axl Rabbit Polyclonal Antibody

NBP1-22964
102160-766EA 536.92 USD
102160-766
Anti-Axl Rabbit Polyclonal Antibody
The Axl Antibody from Novus Biologicals is a rabbit polyclonal antibody to Axl. This antibody reacts with human. The Axl Antibody has been validated for the following applications: Western Blot, Immunoprecipitation (Negative).

Type: Primary
Antigen: Axl
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR