Anti-V1a Vasopressin Receptor Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-44150
102173-884EA 564.74 USD
102173-884
Anti-V1a Vasopressin Receptor Rabbit Polyclonal Antibody
Antibodies
The V1a Vasopressin R / AVPR1A Antibody from Novus Biologicals is a rabbit polyclonal antibody to V1a Vasopressin R / AVPR1A. This antibody reacts with human. The V1a Vasopressin R / AVPR1A Antibody has been validated for the following applications: Immunohistochemistry-Paraffin.

Type: Primary
Antigen: V1a Vasopressin Receptor
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR