Anti-AUTS2 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-32784
102166-384EA 635.51 USD
102166-384
Anti-AUTS2 Rabbit Polyclonal Antibody
Antibodies
The AUTS2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to AUTS2. This antibody reacts with human, mouse. The AUTS2 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: AUTS2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR