Anti-Aurora B Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB100-294
102108-378EA 564.74 USD
102108-378
Anti-Aurora B Rabbit Polyclonal Antibody
Antibodies
The Aurora B Antibody from Novus Biologicals is a rabbit polyclonal antibody to Aurora B. This antibody reacts with human, mouse, rat. The Aurora B Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Microarray.

Type: Primary
Antigen: Aurora B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR