Anti-ATXN7L2 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-70417
102199-604EA 564.74 USD
102199-604
Anti-ATXN7L2 Rabbit Polyclonal Antibody
Antibodies
The ATXN7L2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to ATXN7L2. This antibody reacts with human. The ATXN7L2 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: ATXN7L2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR