Anti-ATP7b Rabbit Polyclonal Antibody (Biotin)

Supplier: Novus Biologicals
NB100-360B
102108-686EA 614.15 USD
102108-686
Anti-ATP7b Rabbit Polyclonal Antibody (Biotin)
Antibodies
The ATP7b Antibody [Biotin] from Novus Biologicals is a rabbit polyclonal antibody to ATP7b. This antibody reacts with human, mouse, rat. The ATP7b Antibody [Biotin] has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: ATP7b
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR