Anti-ATP6V0E2 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-55100
102185-150EA 536.43 USD
102185-150
Anti-ATP6V0E2 Rabbit Polyclonal Antibody
Antibodies
The ATP6V0E2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to ATP6V0E2. This antibody reacts with human. The ATP6V0E2 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: ATP6V0E2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR