Anti-ATP6AP1L Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-91585
102217-708EA 564.74 USD
102217-708
Anti-ATP6AP1L Rabbit Polyclonal Antibody
Antibodies
The ATP6AP1L Antibody from Novus Biologicals is a rabbit polyclonal antibody to ATP6AP1L. This antibody reacts with human. The ATP6AP1L Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: ATP6AP1L
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR