Anti-ATP5E Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Novus Biologicals
NBP1-42805H
102171-650EA 323 USD
102171-650
Anti-ATP5E Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
The ATP5E Antibody [HRP] from Novus Biologicals is a rabbit polyclonal antibody to ATP5E. This antibody reacts with human, mouse, rat, porcine, bovine, xenopus, drosophila (negative). The ATP5E Antibody [HRP] has been validated for the following applications: Western Blot.

Type: Primary
Antigen: ATP5E
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Porcine, Bovine, Xenopus, Drosophila (Negative)
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR