Anti-ATP5E Rabbit Polyclonal Antibody (DyLight 650)

Supplier: Novus Biologicals
NBP1-42813C
102171-664EA 323 USD
102171-664
Anti-ATP5E Rabbit Polyclonal Antibody (DyLight 650)
Antibodies
The ATP5E Antibody [DyLight 650] from Novus Biologicals is a rabbit polyclonal antibody to ATP5E. This antibody reacts with drosophila, human (negative). The ATP5E Antibody [DyLight 650] has been validated for the following applications: Western Blot, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: ATP5E
Clonality: Polyclonal
Clone:
Conjugation: DyLight 650
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Drosophila, Human (Negative)
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR