Anti-ATP5D Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-32365
102165-826EA 635.51 USD
102165-826
Anti-ATP5D Rabbit Polyclonal Antibody
Antibodies
The ATP5D Antibody from Novus Biologicals is a rabbit polyclonal antibody to ATP5D. This antibody reacts with human, mouse. The ATP5D Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: ATP5D
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR