Anti-ATP13A2 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB110-41486 NB110-41486SS
102122-224EA 491.08 USD
102122-224 102122-238
Anti-ATP13A2 Rabbit Polyclonal Antibody
Antibodies
The ATP13A2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to ATP13A2. This antibody reacts with human, mouse. The ATP13A2 Antibody has been validated for the following applications: Western Blot, Simple Western, Immunohistochemistry.

Simple Western,

Type: Primary
Antigen: ATP13A2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR