Anti-NCOA3 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB100-314
102108-500EA 635.51 USD
102108-500
Anti-NCOA3 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal AIB1 Antibody. Tested Applications: Western Blot, Immunoprecipitation, Proximity Ligation Assay. Tested Reactivity: Human.

Type: Primary
Antigen: NCOA3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR