Anti-AGER Rabbit Polyclonal Antibody

NBP1-42612
102171-300EA 423.08 USD
102171-300
Anti-AGER Rabbit Polyclonal Antibody
The AGER Antibody from Novus Biologicals is a rabbit polyclonal antibody to AGER. This antibody reacts with human, mouse, rat. The AGER Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Recommended Dilutions: Immunohistochemistry (Parafin): 0.5-1 ?g/ml

Type: Primary
Antigen: AGER
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR