Anti-AGAP1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-31261
102164-376EA 635.51 USD
102164-376
Anti-AGAP1 Rabbit Polyclonal Antibody
Antibodies
The AGAP1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to AGAP1. This antibody reacts with human, mouse, rat. The AGAP1 Antibody has been validated for the following applications: Western Blot, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: AGAP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR