Anti-AF9 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB100-1565
102105-518EA 635.51 USD
102105-518
Anti-AF9 Rabbit Polyclonal Antibody
Antibodies
The AF9 Antibody from Novus Biologicals is a rabbit polyclonal antibody to AF9. This antibody reacts with human. The AF9 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: AF9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR