Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Adropin Rabbit Polyclonal Antibody (DyLight 550)

Supplier: Novus Biologicals

NBP1-26387R
102160-926EA 429.23 USD InStock
102160-926
Anti-Adropin Rabbit Polyclonal Antibody (DyLight 550)
Antibodies
The Adropin Antibody [DyLight 550] from Novus Biologicals is a rabbit polyclonal antibody to Adropin. This antibody reacts with human, mouse. The Adropin Antibody [DyLight 550] has been validated for the following applications: Western Blot.

Type: Primary
Antigen: Adropin
Clonality: Polyclonal
Clone:
Conjugation: DyLight 550
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty